Sodium 4-aminohippurate hydrate, 98%
Sodium 4-aminohippurate hydrate, 98%
Thermo Scientific Chemicals
Sodium 4-aminohippurate hydrate, 98%
CAS: 206658-83-5 | C9H10N2NaO3+ | 217.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA15775.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium 4-aminohippurate hydrate
CAS206658-83-5
Melting Point123°C to 127°C
Recommended StorageAmbient temperatures
RTECS NumberMR8157200