2-(Ethylamino)ethanol, 98%
2-(Ethylamino)ethanol, 98%
2-(Ethylamino)ethanol, 98%
2-(Ethylamino)ethanol, 98%
Thermo Scientific Chemicals
2-(Ethylamino)ethanol, 98%
CAS: 110-73-6 | C4H11NO | 89.138 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15744.AP500 mL
Catalog number ALFA15744.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material2-(Ethylamino)ethanol
CAS110-73-6
Health Hazard 1H227-H302-H311-H314
Health Hazard 2GHS H Statement
H311-H314-H318-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Health Hazard 3P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.
Solubility
Miscible in water.
Notes
Store at room temperature. Keep away from oxidizing agents and acids.
RUO – Research Use Only
General References:
  1. M. Dupra.; T.; N. Bui.; F. Dabosi. Corrosion Inhibition of a Carbon Steel by 2-Ethylamino-Ethanol in Aerated 3% NaCI Solutions - Effect of pH. Corrosion. 1979, 35 (9), 392-397 .
  2. Estrella Álvarez.; Diego Gómez-Díaz.; M. Dolores La Rubia.; osé M. Navaza. Densities and Viscosities of Aqueous Ternary Mixtures of 2-(Methylamino)ethanol and 2-(Ethylamino)ethanol with Diethanolamine, Triethanolamine, N-Methyldiethanolamine, or 2-Amino-1-methyl-1-propanol from 298.15 to 323.15 K.J. Chem. Eng. Data. 2006, 51 (3),955-962 .