Dextrin, precipitated by alcohol
Dextrin, precipitated by alcohol
Dextrin, precipitated by alcohol
Dextrin, precipitated by alcohol
Thermo Scientific Chemicals
Dextrin, precipitated by alcohol
CAS: 9004-53-9 | C18H32O16 | 504.438 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15717.22100 g
Catalog number ALFA15717.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDextrin
Name Noteprecipitated by alcohol
CAS9004-53-9
Recommended StorageAmbient temperatures
RTECS NumberHH9450000
View more
Dextrin is used as adhesive in the manufacture of gummed tapes, textiles and paper. It is used in producing nutritional products. It is also used as a absorbents, binding agents and viscosity controlling agents.