1-Hexylamine, 99%
1-Hexylamine, 99%
1-Hexylamine, 99%
Thermo Scientific Chemicals
1-Hexylamine, 99%
CAS: 111-26-2 | C6H15N | 101.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15663.AU1000 mL
Catalog number ALFA15663.AU
Price (MYR)
1,606.00
EA
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (MYR)
1,606.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hexylamine
CAS111-26-2
Health Hazard 1H226-H301+H311-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.
Solubility
Miscible with water, dichloromethane, alcohol and acetone.
Notes
Air sensitive. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kizaki, K.; Imoto, H.; Kato, T.; Naka, K. Facile construction of N-alkyl arylaminomaleimide derivatives as intensively emissive aggregation induced emission dyes. Tetrahedron 2015, 71 (4), 643-647.
  2. Navia, P.; Bessieres, D.; Plantier, F. Heat capacity of 1-pentylamine and 1-hexylamine: Experimental determination and modeling through a two-state association model (TSAM). J. Chem. Thermodyn. 2013, 57, 367-371.