4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals
4-Fluorophenylhydrazine hydrochloride, 97%
CAS: 823-85-8 | C6H8ClFN2 | 162.59 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15624.1425 g
Catalog number ALFA15624.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Fluorophenylhydrazine hydrochloride
CAS823-85-8
Health Hazard 1H302+H312+H332-H315-H317-H319-H335-H351
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P362-P501c
View more
4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.
Solubility
Soluble in water.
Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Willard M. Welch; Charles A. Harbert; Albert Weissman. Neuroleptic 4-aryltetrahydropyrrolo[3,4-b]indoles. J. Med. Chem. 1980, 23 (6), 704-707.
  2. Daniele Zampieri; Maria Grazia Mamolo; Erik Laurini; Giuditta Scialino; Elena Banfi; Luciano Vio. Antifungal and antimycobacterial activity of 1-(3,5-diaryl-4,5-dihydro-1H-pyrazol-4-yl)-1H-imidazole derivatives[3,4-b]indoles. Bioorganic & Medicinal Chemistry. 2008, 16 (8), 4516-4522.