Potassium tetrafluoroborate, 98%
Potassium tetrafluoroborate, 98%
Potassium tetrafluoroborate, 98%
Potassium tetrafluoroborate, 98%
Thermo Scientific Chemicals
Potassium tetrafluoroborate, 98%
CAS: 14075-53-7 | BF4K | 125.90 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15605.30250 g
Catalog number ALFA15605.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Boiling Point(decomposition)
Chemical Name or MaterialPotassium tetrafluoroborate
Melting Point529°C
CAS14075-53-7
Health Hazard 1H314
View more
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.
Solubility
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
RUO – Research Use Only
General References:
  1. He, T.; Wang, Y. F.; Zeng, J. H. Stable, High-Efficiency Pyrrolidinium-Based Electrolyte for Solid-State Dye-Sensitized Solar Cells. ACS Appl. Mater. Interfaces 2015, 7 (38), 21381-21390.
  2. Pisarek, B. P.; Rapiejko, C.; Święcik, R.; Pacyniak, T. Effect of Inhibitor Coating of a Ceramic Mould on the Surface Quality of an AM60 Alloy Cast with Cr and V. Arch. Foundry Eng. 2015, 15 (3), 51-56.