Ethyl oleate, tech. 70%
Ethyl oleate, tech. 70%
Ethyl oleate, tech. 70%
Ethyl oleate, tech. 70%
Thermo Scientific Chemicals
Ethyl oleate, tech. 70%
CAS: 111-62-6 | C20H38O2 | 310.522 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15601.30250 g
Catalog number ALFA15601.30
Price (MYR)
340.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
340.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl oleate
CAS111-62-6
Melting Point-32°C
Recommended StorageAmbient temperatures
Density0.87