2-Amino-3-bromo-5-methylbenzoic acid, 98+%
2-Amino-3-bromo-5-methylbenzoic acid, 98+%
2-Amino-3-bromo-5-methylbenzoic acid, 98+%
2-Amino-3-bromo-5-methylbenzoic acid, 98+%
2-Amino-3-bromo-5-methylbenzoic acid, 98+%
CAS: 13091-43-5 | C8H8BrNO2 | 230.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15443.031 g
Catalog number ALFA15443.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-3-bromo-5-methylbenzoic acid
CAS13091-43-5
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c