1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
Thermo Scientific Chemicals
1,1,4,7,7-Pentamethyldiethylenetriamine, 98%
CAS: 3030-47-5 | C9H23N3 | 173.304 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15433.1425 g
Catalog number ALFA15433.14
Price (MYR)
396.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
396.00
EA
Add to cart
Specifications
Chemical Name or Material1,1,4,7,7-Pentamethyldiethylenetriamine
CAS3030-47-5
Health Hazard 1H227-H302-H311-H314-H335
Health Hazard 2GHS H Statement
H311-H314-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Combustible liquid.
Harmful if swallowed.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes
Solubility
Miscible in water.
Notes
Air & Moisture Sensitive. Store under inert gas. Incompatible with strong oxidizing agents, strong acids.
RUO – Research Use Only
General References:
  1. L. Banci; A. Bencini; D. Gatteschi. Anisotropic exchange in transition-metal dinuclear complexes. 3. Bis(.mu.-(1,3-azido)bis(1,1,4,7,7-pentamethyldiethylenetriamine)dicopper(II) bis(tetraphenylborate).Inorg. Chem.1984, 23 (14), 2138-2141.
  2. Hou Chen; Delong Liu; Yuting Song; Rongjun Qu and Chunhua Wang. ARGET ATRP of acrylonitrile with ionic liquid as reaction media and 1,1,4,7,7-pentamethyldiethylenetriamine as both ligand and reducing agent in the presence of air.Polymers for Advanced Technologies.2011, 22 1513-1517.