1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
Thermo Scientific Chemicals
1H-Benzotriazole, 99%
CAS: 95-14-7 | C6H5N3 | 119.127 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15423.30250 g
Catalog number ALFA15423.30
Price (MYR)
657.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
657.00
EA
Add to cart
Specifications
Chemical Name or Material1H-Benzotriazole
CAS95-14-7
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H302-H332-H319
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
View more
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.
Solubility
Soluble in water, benzene, toluene, chloroform, ethanol and N,N-dimethylformamide.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and heavy metals.
RUO – Research Use Only
General References:
  1. The combination with thionyl chloride (1:1), dissolved in DCM, is a useful and convenient reagent for conversion of alcohols and carboxylic acids to the alkyl or acyl chlorides: Synlett, 1763 (1999).
  2. For reviews of Katritzky's extensive work on benzotriazoles as versatile synthetic intermediates, see: Tetrahedron, 47, 2683 (1991); Chem. Soc. Rev., 363 (1994) Synthesis, 445 (1994); 1315 (1995).
  3. For use in the synthesis of indolo[2,3-b]quinolines, see 2-Chloroquinoline, B23443.
  4. Singh, R. K.; Gupta, P.; Sharma, O. P.; Ray, S. S. Homogeneous synthesis of cellulose fatty esters in ionic liquid (1-butyl-3-methylimidazolium chloride) and study of their comparative antifriction property. J. Ind. Eng. Chem. 2015, 24, 14-19.
  5. Terzopoulou, E.; Voutsa, D.; Kaklamanos, G. A multi-residue method for determination of 70 organic micropollutants in surface waters by solid-phase extraction followed by gas chromatography coupled to tandem mass spectrometry. Environ. Sci. Pollut. Res. 2015, 22 (2), 1095-1112.