1-Chloro-2-nitrobenzene, 99%
1-Chloro-2-nitrobenzene, 99%
1-Chloro-2-nitrobenzene, 99%
1-Chloro-2-nitrobenzene, 99%
Thermo Scientific Chemicals
1-Chloro-2-nitrobenzene, 99%
CAS: 88-73-3 | C6H4ClNO2 | 157.55 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15403.30250 g
Catalog number ALFA15403.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Chloro-2-nitrobenzene
CAS88-73-3
Health Hazard 1H302-H311
Health Hazard 2GHS H Statement
H301-H311-H351-H361-H373-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P312-P330-P361-P363-P501c
View more
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.
Solubility
Soluble in water 0.59 g/L.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. I. Mossakowska and G. Wójcik. 1-Chloro-2-nitro­benzene: N-O...Cl halogen bonds and aromatic [pi]-[pi] stacking, and thermal vibrations in the vicinity of the melting point. Acta Cryst. 2007, c63, o123-o125.
  2. Robert Gatermann; Heinrich Hühnerfuss; Gerhard Rimkus; Manfred Wolf; Stephan Franke. The distribution of nitrobenzene and other nitroaromatic compounds in the North Sea. Marine Pollution Bulletin. 1995, 30(3), 221-227.