1,6-Diiodohexane, 97+%, stab. with copper
1,6-Diiodohexane, 97+%, stab. with copper
1,6-Diiodohexane, 97+%, stab. with copper
1,6-Diiodohexane, 97+%, stab. with copper
Thermo Scientific Chemicals
1,6-Diiodohexane, 97+%, stab. with copper
CAS: 629-09-4 | C6H12I2 | 337.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA15368.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1,6-Diiodohexane
Name Notestabilized with copper
CAS629-09-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
View more
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Page, Z. A.; Liu, F.; Russell, T. P.; Emrick, T. Tuning the energy gap of conjugated polymer zwitterions for efficient interlayers and solar cells. J. Polym. Sci., Part A: Polym. Chem. 2015, 53 (2), 327-336.
  2. Lee, M.; Choi, U. H.; Colby, R. H.; Gibson, H. W. Ion Conduction in a Semicrystalline Polyviologen and Its Polyether Mixtures. Macromol. Chem. Phys. 2015, 216 (3), 344-349.