2-Iodo-1,3,5-trimethylbenzene, 98%
2-Iodo-1,3,5-trimethylbenzene, 98%
2-Iodo-1,3,5-trimethylbenzene, 98%
2-Iodo-1,3,5-trimethylbenzene, 98%
Thermo Scientific Chemicals
2-Iodo-1,3,5-trimethylbenzene, 98%
CAS: 4028-63-1 | C9H11I | 246.091 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15360.1425 g
Catalog number ALFA15360.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Iodo-1,3,5-trimethylbenzene
CAS4028-63-1
Melting Point28°C to 30°C
Recommended StorageAmbient temperatures
EINECS Number223-709-9