2-Bromo-4'-fluoroacetophenone, 97%
2-Bromo-4'-fluoroacetophenone, 97%
2-Bromo-4'-fluoroacetophenone, 97%
2-Bromo-4'-fluoroacetophenone, 97%
Thermo Scientific Chemicals
2-Bromo-4'-fluoroacetophenone, 97%
CAS: 403-29-2 | C8H6BrFO | 217.037 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15319.065 g
Catalog number ALFA15319.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-4'-fluoroacetophenone
CAS403-29-2
Health Hazard 1H314
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
2-Bromo-4'-fluoroacetophenone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. David B.Reitz; James J.Li; Monica B.Norton; Emily J.Reinhard; Joe T.Collins; Gary D.Anderson; Susan A.Gregory; Carol M.Koboldt; William E.Perkins. Selective cyclooxygenase inhibitors: Novel 1,2-diarylcyclopentenes are potent and orally active COX-2 inhibitors. J. Med. Chem. 1994, 37,(23), 3878-3881.
  2. Cheng-Chyi Tzeng; Kuan-Han Lee; Tai-Chi Wang; Chein-Hwa Han; Yeh-Long Chen. Synthesis and cytotoxic evaluation of a series of γ-substituted γ-aryloxymethyl-α-methylene-γ-butyrolactones against cancer cells. Pharmaceutical Research. 2000, 17,(6), 715-719.