4'-Hydroxypropiophenone, 99%
4'-Hydroxypropiophenone, 99%
4'-Hydroxypropiophenone, 99%
4'-Hydroxypropiophenone, 99%
Thermo Scientific Chemicals
4'-Hydroxypropiophenone, 99%
CAS: 70-70-2 | C9H10O2 | 150.177 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15314.22100 g
Catalog number ALFA15314.22
Price (MYR)
374.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
374.00
EA
Add to cart
Specifications
Chemical Name or Material4'-Hydroxypropiophenone
CAS70-70-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c