4-Hydroxychalcone, 97%
4-Hydroxychalcone, 97%
4-Hydroxychalcone, 97%
4-Hydroxychalcone, 97%
Thermo Scientific Chemicals
4-Hydroxychalcone, 97%
CAS: 20426-12-4 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA15307.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Hydroxychalcone
CAS20426-12-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. O.A. Oyedapo; C.O. Adewunmi; E.O. Iwalewa and V.O. Makanju. Analgesic, Antioxidant and Anti-inflammatory Related Activities of 21-hydroxy-2,41-dimethoxychalcone and 4-hydroxychalcone in Mice.Journal of Biological Sciences.2008, 8 131-136.
  2. Barbora Orlikovaa; Deniz Tasdemirb; Frantisek Golaisc; Mario Dicatoa; Marc Diedericha. The aromatic ketone 4'-hydroxychalcone inhibits TNFα-induced NF-κB activation via proteasome inhibition.Biochemical Pharmacology.2011, 82 620-631.