Metaldehyde, 98%
Metaldehyde, 98%
Metaldehyde, 98%
Thermo Scientific Chemicals
Metaldehyde, 98%
CAS: 108-62-3 | C8H16O4 | 176.212 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15292.1850 g
Catalog number ALFA15292.18
Price (MYR)
308.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
308.00
EA
Add to cart
Specifications
Chemical Name or MaterialMetaldehyde
CAS108-62-3
Health Hazard 1H228-H301-H330
Health Hazard 2GHS H Statement
H301-H330-H228
Toxic if swallowed.
Fatal if inhaled.
Flammable solid.
Health Hazard 3P210-P240-P241-P260-P264b-P270-P271-P280-P284-P301+P310-P304+P340-P310-P314-P330-P370+P378q-P501c