Linoleic acid, tech. 75%
Linoleic acid, tech. 75%
Linoleic acid, tech. 75%
Linoleic acid, tech. 75%
Linoleic acid, tech. 75%
CAS: 60-33-3 | C18H32O2 | 280.452 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15265.1850 g
Catalog number ALFA15265.18
Price (MYR)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLinoleic acid
CAS60-33-3
Recommended StorageKeep cold
Density0.902
RTECS NumberRF9990000