Indole-3-butyric acid, 98%
Indole-3-butyric acid, 98%
Indole-3-butyric acid, 98%
Indole-3-butyric acid, 98%
Thermo Scientific Chemicals
Indole-3-butyric acid, 98%
CAS: 133-32-4 | C12H13NO2 | 203.241 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15260.065 g
Catalog number ALFA15260.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIndole-3-butyric Acid
CAS133-32-4
Health Hazard 1H301-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.
Solubility
Soluble in water(0.25g/L).
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Lucia C. Strader.; Bonnie Bartel. Transport and Metabolism of the Endogenous Auxin Precursor Indole-3-Butyric Acid. Molecular Plant. 2011, 4, (3), 477-486.
  2. Jutta Ludwig-Muller. Indole-3-butyric acid in plant growth and development. Plant Growth Regulation. 2000, 32, (2), 219-230.