4-Acetylbenzoic acid, 98+%
4-Acetylbenzoic acid, 98+%
4-Acetylbenzoic acid, 98+%
4-Acetylbenzoic acid, 98+%
Thermo Scientific Chemicals
4-Acetylbenzoic acid, 98+%
CAS: 586-89-0 | C9H8O3 | 164.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15239.1425 g
Catalog number ALFA15239.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Acetylbenzoic acid
CAS586-89-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Paula Rodrigues; Karen Scheuermann; Cornelia Stockmar; Gerhard Maier; Heinz Fiebig; Clemens Unger; Rolf Mulhaupt; Felix Kratz. Synthesis and in vitro efficacy of acid-sensitive poly(ethylene glycol) paclitaxel conjugates. Bioorganic & Medicinal Chemistry Letters. 2003, 13,(3), 355-360.
  2. Niti Sunsandee; Milan Hronec; Magdalena Stolcova; Natchanun Leepipatpiboon; Ura Pancharoen. Thermodynamics of the solubility of 4-acetylbenzoic acid in different solvents from 303.15 to 473.15 K. Journal of Molecular Liquids. 2013, 180,252-259.