1,3-Diphenylguanidine, 97%
1,3-Diphenylguanidine, 97%
1,3-Diphenylguanidine, 97%
1,3-Diphenylguanidine, 97%
Thermo Scientific Chemicals
1,3-Diphenylguanidine, 97%
CAS: 102-06-7 | C13H13N3 | 211.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1000 g
Catalog number ALFA15234.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Diphenylguanidine
CAS102-06-7
Health Hazard 1H302-H315-H319-H335-H361f
Health Hazard 2GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
1,3-Diphenylguanidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Diphenylguanidine is used as pharmaceutical intermediate.
Solubility
Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. P.V.Shah; M.R.Sumler; Y.M.Ioannou; H.L.Fisher; L.L.Hall. Dermal absorption and disposition of 1,3-diphenylguanidine in rats. Journal of Toxicology and Environmental Health. 1985, 15,(5), 623-633.
  2. P.V.Gushchin; M.Haukka; N.A.Bokach; V.Yu.Kukushkin. Platinum(iv)-mediated nucleophilic addition of 1,3-diphenylguanidine to propiononitrile. Russian Chemical Bulletin. 2008, 57,(10), 2125-2131.