2,5-Dimethylhexane, 99%
2,5-Dimethylhexane, 99%
2,5-Dimethylhexane, 99%
2,5-Dimethylhexane, 99%
Thermo Scientific Chemicals
2,5-Dimethylhexane, 99%
CAS: 592-13-2 | C8H18 | 114.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15221.065 g
Catalog number ALFA15221.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2,5-Dimethylhexane
CAS592-13-2
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 2GHS H Statement
H304-H226-H315-H336
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c