2,4-Difluorobenzyl bromide, 98+%
2,4-Difluorobenzyl bromide, 98+%
2,4-Difluorobenzyl bromide, 98+%
2,4-Difluorobenzyl bromide, 98+%
Thermo Scientific Chemicals
2,4-Difluorobenzyl bromide, 98+%
CAS: 23915-07-3 | C7H5BrF2 | 207.018 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15213.065 g
Catalog number ALFA15213.06
Price (MYR)
374.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
374.00
EA
Add to cart
Specifications
Chemical Name or Material2,4-Difluorobenzyl bromide
CAS23915-07-3
Health Hazard 1H226-H314
Health Hazard 2GHS H Statement
H314-H318-H317-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Health Hazard 3P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c