Tetrahydrofurfuryl bromide, 95%
Tetrahydrofurfuryl bromide, 95%
Tetrahydrofurfuryl bromide, 95%
Tetrahydrofurfuryl bromide, 95%
Thermo Scientific Chemicals
Tetrahydrofurfuryl bromide, 95%
CAS: 1192-30-9 | C5H9BrO | 165.03 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15192.065 g
Catalog number ALFA15192.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydrofurfuryl bromide
CAS1192-30-9
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c