2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA15134.22100 g
Catalog number ALFA15134.22
Price (MYR)
277.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
277.00
EA
Add to cart
Specifications
Chemical Name or Material2-Dimethylaminoethyl chloride hydrochloride
CAS4584-46-7
Health Hazard 1H301-H311-H315-H320-H335-H341-H372
Health Hazard 2GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
View more
2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.
RUO – Research Use Only
General References:
  1. Ali A. El-Emam; Omar A. Al-Deeb; Mohamed Al-Omar; Jochen Lehmann. Synthesis, antimicrobial, and anti-HIV-1 activity of certain 5-(1-adamantyl)-2-substituted thio-1,3,4-oxadiazoles and 5-(1-adamantyl)-3-substituted aminomethyl-1,3,4-oxadiazoline-2-thiones. Bioorganic & Medicinal Chemistry., 2004, 12(19), 5107-5113.
  2. Harvey Gurien. Reaction of 2-Dimethylaminoethyl Chloride with Magnesium. J. Org. Chem., 1963, 28(3), 878-879.