3-Methoxyphenyl isothiocyanate, 98%
3-Methoxyphenyl isothiocyanate, 98%
3-Methoxyphenyl isothiocyanate, 98%
3-Methoxyphenyl isothiocyanate, 98%
Thermo Scientific Chemicals
3-Methoxyphenyl isothiocyanate, 98%
CAS: 3125-64-2 | C8H7NOS | 165.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15090.065 g
Catalog number ALFA15090.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Methoxyphenyl isothiocyanate
CAS3125-64-2
Health Hazard 1H302+H312+H332-H314
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c