Tri-n-octylamine, 95%
Tri-n-octylamine, 95%
Tri-n-octylamine, 95%
Tri-n-octylamine, 95%
Thermo Scientific Chemicals
Tri-n-octylamine, 95%
CAS: 1116-76-3 | C24H51N | 353.679 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15067.AE100 mL
Catalog number ALFA15067.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-octylamine
CAS1116-76-3
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, carbon dioxide, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only
General References:
  1. Datta, D.; Uslu, H.; Kumar, S. Intensification of picolinic acid extraction with tri-n-butylphosphate and tri-n-octylamine in three different diluents. Chem. Eng. Res. Des. 2015, 95, 105-112.
  2. Pal, D.; Tripathi, A.; Shukla, A.; Gupta, K. R.; Keshav, A. Reactive extraction of pyruvic acid using tri-n-octylamine diluted in decanol/kerosene: equilibrium and effect of temperature. J. Chem. Eng. Data 2015, 60 (3), 860-869.