3,4,5-Trimethoxybenzaldehyde, 98%
3,4,5-Trimethoxybenzaldehyde, 98%
3,4,5-Trimethoxybenzaldehyde, 98%
3,4,5-Trimethoxybenzaldehyde, 98%
Thermo Scientific Chemicals
3,4,5-Trimethoxybenzaldehyde, 98%
CAS: 86-81-7 | C10H12O4 | 196.202 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15038.1425 g
Catalog number ALFA15038.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3,4,5-Trimethoxybenzaldehyde
CAS86-81-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Melting Point73°C to 77°C
View more
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.
Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. D.V Rao: F.A. Stuber. An efficient synthesis of 3, 4, 5-trimethoxybenzaldehyde from vanillin. Synthesis, 2013, 4 680-689.
  2. Percy S. Manchand; Peter S. Belica; Harry S. Wonga. Synthesis of 3,4,5-Trimethoxybenzaldehyde. Talanta. 1990, 20(17),2659-2666.