L-(+)-Asparagine monohydrate, 98+%
L-(+)-Asparagine monohydrate, 98+%
L-(+)-Asparagine monohydrate, 98+%
L-(+)-Asparagine monohydrate, 98+%
Thermo Scientific Chemicals
L-(+)-Asparagine monohydrate, 98+%
CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15012.22100 g
Catalog number ALFA15012.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-(+)-Asparagine monohydrate
Melting Point∼245°C (decomposition)
CAS5794-13-8
Recommended StorageAmbient temperatures
EINECS Number200-735-9
View more
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Starting material for synthesis of ß -amino acids: Arch. Pharm., 324, 551 (1991)
  2. Shkir, M.; Muhammad, S.; AlFaify, S.; Irfan, A.; Yahia, I. S. A dual approach to study the electro-optical properties of a noncentrosymmetric L-asparagine monohydrate. Spectrochim. Acta, Part A 2015, 137, 432-441.
  3. Thukral, K.; Vijayan, N.; Haranath, D.; Jayaramakrishnan, V.; Philip, J.; Sreekanth, P.; Bhagavannaryana, G. Assessment on third order non linearity and other optical analyses of L-Asparagine Monohydrate single crystal: An efficient candidate for harmonic conversions. Spectrochim. Acta, Part A 2015, 151, 419-425.