1-Dodecene, 96%
1-Dodecene, 96%
1-Dodecene, 96%
Thermo Scientific Chemicals
1-Dodecene, 96%
CAS: 112-41-4 | C12H24 | 168.324 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14992.22100 g
Catalog number ALFA14992.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Dodecene
CAS112-41-4
Health Hazard 1H227-H304
Health Hazard 2GHS H Statement
H304-H315-H227
May be fatal if swallowed and enters airways.
Causes skin irritation.
Combustible liquid.
Health Hazard 3P210-P280-P301+P310-P331-P370+P378q-P501c
View more
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.
Solubility
It is soluble in ethanol, ethyl ether, and acetone
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
RUO – Research Use Only
General References:
  1. Kazuhiko Sato.; Masao Aoki.; Masami Ogawa.; Tadashi Hashimoto.; Ryoji Noyori. A Practical Method for Epoxidation of Terminal Olefins with 30% Hydrogen Peroxide under Halide-Free Conditions. J. Org. Chem. 1996, 61, (23), 8310-8311.
  2. Christine E. Garrett.; Gregory C. Fu. Hydroboration of Olefins with Catecholborane at Room Temperature in the Presence of N,N-Dimethylacetamide. J. Org. Chem. 1996, 61, (10), 3224-3225.