2-Fluorophenol, 98%
2-Fluorophenol, 98%
2-Fluorophenol, 98%
Thermo Scientific Chemicals
2-Fluorophenol, 98%
CAS: 367-12-4 | C6H5FO | 112.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14979.1425 g
Catalog number ALFA14979.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Fluorophenol
CAS367-12-4
Health Hazard 1H226-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H311-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c