2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™
2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™
2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™
2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™
2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA14935.065 g
Catalog number ALFA14935.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Nitrophenyl beta-D-glucopyranoside
CAS2816-24-2
Melting Point141°C to 142°C
Recommended StorageKeep cold
EINECS Number220-568-5
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Nitrophenyl beta-D-glucopyranoside is used as a substrate for ß -glucosidase.
Solubility
Soluble in water
Notes
Keep container tightly closed. Store away from oxidizing agents.
General References:
  1. Substrate for ß -glucosidase.