Terephthalaldehyde, 98%
Terephthalaldehyde, 98%
Terephthalaldehyde, 98%
Terephthalaldehyde, 98%
Thermo Scientific Chemicals
Terephthalaldehyde, 98%
CAS: 623-27-8 | C8H6O2 | 134.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA14930.22
Price (MYR)
572.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
572.00
EA
Add to cart
Specifications
Chemical Name or MaterialTerephthalaldehyde
CAS623-27-8
Health Hazard 1H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
View more
Terephthalaldehyde is used as an intermediate for the preparation of dyes and fluorescent whitening agents. It is used in the synthesis of polyimines by reacting with aliphatic diamines. It is used as a precursor for the preparation of paramagnetic microporous polymeric organic frameworks (POFs) through copolymerization with pyrrole, indole, and carbazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Terephthalaldehyde is used as an intermediate for the preparation of dyes and fluorescent whitening agents. It is used in the synthesis of polyimines by reacting with aliphatic diamines. It is used as a precursor for the preparation of paramagnetic microporous polymeric organic frameworks (POFs) through copolymerization with pyrrole, indole, and carbazole.
Solubility
Soluble in hot water, alcohol and ether.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Xu, B.; Zheng, D.; Qiu, W.; Gao, F.; Jiang, S.; Wang, Q. An ultrasensitive DNA biosensor based on covalent immobilization of probe DNA on fern leaf-like alfa-Fe2O3 and chitosan Hybrid film using terephthalaldehyde as arm-linker. Biosens. Bioelectron. 2015, 72, 175-181.
  2. Rohan, R.; Pareek, K.; Cai, W.; Zhang, Y.; Xu, G.; Chen, Z.; Gao, Z.; Danb, Z.; Cheng, H. Melamine-terephthalaldehyde-lithium complex: a porous organic network based single ion electrolyte for lithium ion batteries. J. Mater. Chem. A 2015, 3 (9), 5132-5139.