2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
Thermo Scientific Chemicals
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%
CAS: 7128-64-5 | C26H26N2O2S | 430.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA14928.22
Price (MYR)
2,346.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,346.00
EA
Add to cart
Specifications
Chemical Name or Material2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
CAS7128-64-5
Melting Point200°C to 202°C
Recommended StorageAmbient temperatures
RTECS NumberDM4888332
View more
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.
Solubility
Soluble in acetone and toluene. Insoluble in water.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Fourati, M. A.; Maris, T.; Skene, W. G.; Bazuin, C G.; Prud’homme, R. E. Photophysical, Electrochemical and Crystallographic Investigations of the Fluorophore 2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene. J. Phys. Chem. B 2011, 115 (43), 12362-12369.
  2. Giani, M.; Sist, P.; Berto, D.; Serrazanetti, G. P.; Ventrella, V.; Urbani, R. The organic matrix of pelagic mucilaginous aggregates in the Tyrrhenian Sea (Mediterranean Sea). Mar. Chem. 2012, 132, 83-94.