Fluorene-2-carboxaldehyde, 99%
Fluorene-2-carboxaldehyde, 99%
Fluorene-2-carboxaldehyde, 99%
Fluorene-2-carboxaldehyde, 99%
Thermo Scientific Chemicals
Fluorene-2-carboxaldehyde, 99%
CAS: 30084-90-3 | C14H10O | 194.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14911.1425 g
Catalog number ALFA14911.14
Price (MYR)
2,357.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,357.00
EA
Add to cart
Specifications
Chemical Name or MaterialFluorene-2-carboxaldehyde
CAS30084-90-3
Health Hazard 1H500
Melting Point82°C to 86°C
RTECS NumberLL5917000