4-Bromophenol, 99%
4-Bromophenol, 99%
4-Bromophenol, 99%
Thermo Scientific Chemicals
4-Bromophenol, 99%
CAS: 106-41-2 | C6H5BrO | 173.009 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14873.22100 g
Catalog number ALFA14873.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromophenol
CAS106-41-2
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H312-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.
Solubility
It is insoluble in water. It is soluble in 5%ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Air sensitive. Keep away from Oxidizing agents.
RUO – Research Use Only
N.W. Larsen. Microwave spectra and internal rotation of 4-fluorophenol, 4-chlorophenol and 4-bromophenol. Journal of Molecular Structure. 1986, 144, (1), 83-99.