2-Hexanone, 98%
2-Hexanone, 98%
2-Hexanone, 98%
Thermo Scientific Chemicals
2-Hexanone, 98%
CAS: 591-78-6 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
50 g
100 g
Catalog number ALFA14867.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Hexanone
CAS591-78-6
Health Hazard 1H226-H336-H361f-H372
Health Hazard 2GHS H Statement
H372-H361-H226-H336
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapour.
May cause drowsiness or dizziness.
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c
View more
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (±)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (+/-)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.
Solubility
Miscible with acetone, ethanol and ether. Slightly miscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases and reducing agents.
RUO – Research Use Only
General References:
  1. Kang, J.; Vonderheide, A.; Guliants, V. V. Deuterium-Labeling Study of the Hydrogenation of 2-Methylfuran and 2,5-Dimethylfuran over Carbon-Supported Noble Metal Catalysts. ChemSusChem 2015, 8 (18), 3044-3047.
  2. Wu, Y. Y.; Yang, C. X.; Yan, X. P. An in situ growth approach to the fabrication of zeolite imidazolate framework-90 bonded capillary column for gas chromatography separation. Analyst 2015, 140 (9), 3107-3112.