Pyrrolidine, 99%
Pyrrolidine, 99%
Pyrrolidine, 99%
Thermo Scientific Chemicals
Pyrrolidine, 99%
CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFA14852.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPyrrolidine
CAS123-75-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H225-H301-H314-H318-H332
Health Hazard 3GHS P Statement
P210-P260-P261-P280-P301+P310-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
View more
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.
Solubility
Miscible with alcohol, ether, chloroform and water.
Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
RUO – Research Use Only
General References:
  1. Pohl, R.; Slavětínská, L. P.; Eng, W. S.; Keough, D. T.; Guddat, L. W.; Rejman, D. Synthesis, conformational studies, and biological properties of phosphonomethoxyethyl derivatives of nucleobases with a locked conformation via a pyrrolidine ring. Org. Biomol. Chem. 2015, 13, 4693-4705.
  2. Redondo, J. A.; Velasco, D.; Pérez-Perrino, M.; Reinecke, H.; Gallardo, A.; Pandit, A.; Elvira, C. Synergistic effect of pendant hydroxypropyl and pyrrolidine moieties randomly distributed along polymethacrylamide backbones on in vitro DNA-transfection. Eur. J. Pharm. Biopharm. 2015, 90, 38-43.