2-Amino-4-methylthiazole, 98+%
2-Amino-4-methylthiazole, 98+%
2-Amino-4-methylthiazole, 98+%
2-Amino-4-methylthiazole, 98+%
Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%
CAS: 1603-91-4 | C4H6N2S | 114.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA14847.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-4-methylthiazole
CAS1603-91-4
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c