Isophorone, 97%
Isophorone, 97%
Isophorone, 97%
Thermo Scientific Chemicals
Isophorone, 97%
CAS: 78-59-1 | C9H14O | 138.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14844.AK250 mL
Catalog number ALFA14844.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialIsophorone
CAS78-59-1
Health Hazard 1H227-H302+H312-H319-H335-H351
Health Hazard 2GHS H Statement
H351-H302-H312-H319-H335-H227
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P201-P202-P210-P235-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P370+P378q-P501c
View more
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings
Solubility
Soluble in water (12g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. M. S. Kharasch.; P. O. Tawney. Factors Determining the Course and Mechanisms of Grignard Reactions. II. The Effect of Metallic Compounds on the Reaction between Isophorone and Methylmagnesium Bromide. J. Am. Chem. Soc. 1941, 63, (9), 2308-2316.
  2. N.S. Zarghami.; D.E. Heinz. Monoterpene aldehydes and isophorone-related compounds of saffron. Phytochemistry. 1971, 10, (11), 2755-2761.