5-Bromouracil, 98+%
5-Bromouracil, 98+%
5-Bromouracil, 98+%
Thermo Scientific Chemicals
5-Bromouracil, 98+%
CAS: 51-20-7 | C4H3BrN2O2 | 190.98 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14799.1425 g
Catalog number ALFA14799.14
Price (MYR)
541.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
541.00
EA
Add to cart
Specifications
Chemical Name or Material5-Bromouracil
CAS51-20-7
Health Hazard 1H302+H312+H332-H315-H319-H335-H341
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c