2-Picolinic acid, 99%
2-Picolinic acid, 99%
2-Picolinic acid, 99%
2-Picolinic acid, 99%
Thermo Scientific Chemicals
2-Picolinic acid, 99%
CAS: 98-98-6 | C6H5NO2 | 123.111 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14749.22100 g
Catalog number ALFA14749.22
Price (MYR)
435.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
435.00
EA
Add to cart
Specifications
Chemical Name or Material2-Picolinic acid
CAS98-98-6
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
View more
2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.
Solubility
Soluble in water. Also soluble in glacial acetic acid, ethanol and methanol. Insoluble in chloroform.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents and bases.
RUO – Research Use Only
General References:
  1. Hong Zhao; Yuzhong Zhang; Zhuobin Yuan. Electrochemical determination of dopamine using a poly(2-picolinicacid) modified glassy carbon electrode. Analyst. 2001, 126, (3),358-360
  2. Xinjian Li; Dapeng Zou; Faqiang Leng; Chunxia Sun; Jingya Li; Yangjie Wu; Yusheng Wu. Arylation of 2-substituted pyridines via Pd-catalyzed decarboxylative cross-coupling reactions of 2-picolinic acid. Chemical Communications. 2013, 49, (3),312-314
  3. Has been found to accelerate significantly oxidations with chromic acid: J. Am. Chem. Soc., 98, 1026 (1976).
  4. Amino groups can be protected, by DCC coupling, as picolinamides, which can be cleaved with acidic Cu(OAc)2: Synth. Commun., 383 (1972).