L-Arginine monohydrochloride, 98+%
L-Arginine monohydrochloride, 98+%
L-Arginine monohydrochloride, 98+%
L-Arginine monohydrochloride, 98+%
Thermo Scientific Chemicals
L-Arginine monohydrochloride, 98+%
L-Arginine monohydrochloride (L-Arginine hydrochloride or arginine HCl, CAS # 1119-34-2) is the salt of the essential amino acid arginine. | CAS: 1119-34-2 | C6H15ClN4O2 | 210.662 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14730.22100 g
Catalog number ALFA14730.22
Price (MYR)
346.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
346.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Arginine monohydrochloride
Melting Point216°C (decomposition)
CAS1119-34-2
Recommended StorageAmbient temperatures
RTECS NumberCF1995500
View more
L-Arginine monohydrochloride is a precursor of nitric oxide synthesis that induces vasodilation, there by relaxing blood vessels. It is an important intermediate in the urea cycle and involved in the detoxification of nitrogenous wastes. It is commonly used in cell culture media and drug development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • L-Arginine is the left-handed enantiomer of the amino acid arginine, an essential amino acid in juvenile humans
  • Arginine residues are common active site components in enzymes
  • Arginine is involved in nitric acid production, and many biological processes depend on it
  • Arginine plays a role in the immune system and in the detoxification of nitrogenous wastes
Application
  • L-Arginine monohydrochloride is suitable for biochemistry and in vitro research
RUO – Research Use Only
General References:
  1. Sangeetha, K.; Babu, R. R.; Kumar, P.; Bhagvannarayana, G.; Ramamurthi, K. Effect of H+ ion implantation on structural, morphological, optical and dielectric properties of l-arginine monohydrochloride monohydrate single crystals. Appl. Surf. Sci. 2011, 257 (17), 7573-7578.
  2. Sangeetha, K.; Babu, R. R.; Bhagavannarayana, G.; Ramamurthi, K. Unidirectional growth and characterization of L-arginine monohydrochloride monohydrate single crystals. Mater. Chem. Phys. 2011, 130 (1), 487-492.