1-Bromooctane, 98+%
1-Bromooctane, 98+%
1-Bromooctane, 98+%
Thermo Scientific Chemicals
1-Bromooctane, 98+%
CAS: 111-83-1 | C8H17Br | 193.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14722.30250 g
Catalog number ALFA14722.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromooctane
CAS111-83-1
Health Hazard 1H227
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only
General References:
  1. Iqbal, Z.; Wu, W. Q.; Huang, Z. S.; Wang, L.; Kuang, D. B.; Meier, H.; Cao, D. Trilateral pi-conjugation extensions of phenothiazine-based dyes enhance the photovoltaic performance of the dye-sensitized solar cells. Dyes Pigm. 2016, 124, 63-71.
  2. Sun, T.; Qi, M.; Fu, R. Perhydroxylcucurbit[6]uril as a highly selective gas chromatographic stationary phase for analytes of wide-ranging polarity. J. Sep. Sci. 2015, 38 (5), 821-824.