2-Iodoacetamide, 98%, stab. with ca 5-8% water
2-Iodoacetamide, 98%, stab. with ca 5-8% water
2-Iodoacetamide, 98%, stab. with ca 5-8% water
2-Iodoacetamide, 98%, stab. with ca 5-8% water
Thermo Scientific Chemicals
2-Iodoacetamide, 98%, stab. with ca 5-8% water
CAS: 144-48-9 | C2H4INO | 184.964 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA14715.14
Price (MYR)
632.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
632.00
EA
Add to cart
Specifications
Chemical Name or MaterialIodoacetamide
Name Notestabilized with ≈5 to 8% water
CAS144-48-9
Health Hazard 1H301-H314-H317-H334-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H317-H335
View more
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.
Solubility
Soluble in water, dimethyl formamide and ethanol. Slightly soluble in methanol.
Notes
Store in a cool place. Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Zbancioc, G.; Mangalagiu, I. I.; Moldoveanu, C. Ultrasound assisted synthesis of imidazolium salts: An efficient way to ionic liquids. Ultrason. Sonochem. 2015, 23, 376-384.
  2. Moriyama, M.; Metzger, S.; van der Vlies, A. J.; Uyama, H.; Ehrbar, M.; Hasegawa, U. Inhibition of Angiogenesis by Antioxidant Micelles. Adv. Healthcare Mater. 2013, 4 (4), 569-575.