2'-Chloroacetanilide, 98+%
2'-Chloroacetanilide, 98+%
2'-Chloroacetanilide, 98+%
2'-Chloroacetanilide, 98+%
Thermo Scientific Chemicals
2'-Chloroacetanilide, 98+%
CAS: 533-17-5 | C8H8ClNO | 169.61 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14711.22100 g
Catalog number ALFA14711.22
Price (MYR)
593.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
593.00
EA
Add to cart
Specifications
Chemical Name or Material2'-Chloroacetanilide
Melting Point86°C to 88°C
CAS533-17-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement