3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
Thermo Scientific Chemicals
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%
CAS: 88-26-6 | C15H24O2 | 236.355 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14709.1425 g
Catalog number ALFA14709.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
CAS88-26-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c