Di-tert-butyl dicarbonate, 97+%
Di-tert-butyl dicarbonate, 97+%
Di-tert-butyl dicarbonate, 97+%
Di-tert-butyl dicarbonate, 97+%
Thermo Scientific Chemicals
Di-tert-butyl dicarbonate, 97+%
CAS: 24424-99-5 | C10H18O5 | 218.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA14708.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-tert-butyl Dicarbonate
CAS24424-99-5
Health Hazard 1H228-H315-H317-H318-H330-H335-H500
Health Hazard 2GHS H Statement
H330-H318-H228-H315-H317-H335
Health Hazard 3P210-P240-P241-P260-P264b-P271-P272-P280-P284-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P370+P378q-P501c
View more
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.
Solubility
Miscible with decalin, toluene, carbon tetrachloride, tetrahydrofuran, dioxane, alcohols, acetone, acetonitrile and dimethylformamide. Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Moisture and light sensitive. Incompatible with oxidizing agents, reducing agents, bases and acids.
RUO – Research Use Only
General References:
  1. Reagent for amino group protection as the tert-butyl carbamate (tert-butoxycarbonyl, Boc derivatives), in high yield under mild conditions, widely used in peptide chemistry; see Appendix 6. For practical details, literature references and tabulated results, see: Org. Synth. Coll., 7, 70 (1990). For further examples see: Synthesis, 223 (1987): Org. Synth. Coll., 9, 124, 300 (1998). Benzyl carbamates (Cbz, Z) may be transformed into Boc in a one pot procedure catalyzed by Pd/C: Tetrahedron Lett., 33, 3167 (1992).
  2. Rapid and efficient formation of Boc derivatives of amines, catayzed by Copper(II) tetrafluoroborate hexahydrate, 26127 has been described: Tetrahdron Lett., 47, 1087 (2006).
  3. For Boc protection of phenols, alcohols, thiols etc. under phase-transfer conditions, see: Can. J. Chem., 63, 153 (1985).
  4. Amides can be protected in the presence of a catalytic quantity of DMAP ( 4-(Dimethyl amino) pyridine, A13016 ): J. Org. Chem., 48, 2424 (1983); Acta Chem. Scand. B, 40, 745 (1986); the amide link of the product undergoes facile alkaline hydrolysis or methanolysis. The nitrogen function of indoles and pyrroles can also be protected under similar conditions: J. Chem. Soc., Chem. Commun., 1699 (1984); Org. Synth. Coll., 9, 121 (1998). 1-Boc indoles may be synthesized from N-Boc 2-alkylanilines: Synthesis, 871 (1991).