n-Tetradecane, 99%
n-Tetradecane, 99%
n-Tetradecane, 99%
n-Tetradecane, 99%
CAS: 629-59-4 | C14H30 | 198.394 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14672.22100 g
Catalog number ALFA14672.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Tetradecane
CAS629-59-4
Health Hazard 1H304
Health Hazard 2GHS H Statement
H304-H336
Health Hazard 3P301+P310-P331-P501c
View more
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Brown, P. S.; Bhushan, B. Mechanically durable, superomniphobic coatings prepared by layer-by-layer technique for self-cleaning and anti-smudge. J. Colloid Interface Sci. 2015, 456, 210-218.
  2. Fan, D.; Zheng, P.; Ma, Y.; Yin, T.; Zhao, J.; Shen, W. Effects of water content and chain length of n-alkane on the interaction enthalpy between the droplets in water/sodium bis (2-ethylhexyl)-sulfosuccinate-alkane microemulsions. Soft Matter 2015, 11 (14), 2885-2892.