Diethylene glycol monoethyl ether, 98%
Diethylene glycol monoethyl ether, 98%
Diethylene glycol monoethyl ether, 98%
Diethylene glycol monoethyl ether, 98%
Thermo Scientific Chemicals
Diethylene glycol monoethyl ether, 98%
CAS: 111-90-0 | C6H14O3 | 134.175 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14670.0C10,000 g
Catalog number ALFA14670.0C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10,000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylene glycol monoethyl ether
CAS111-90-0
Health Hazard 1H319
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.
Solubility
Fully miscible in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. A.Ganem-Quintanar; C.Lafforgue; F.Falson-Rieg; P.Buri. Evaluation of the transepidermal permeation of diethylene glycol monoethyl ether and skin water loss. International Journal of Pharmaceutics. 1997, 147, (2),165-171
  2. M.Yazdanian; E.Chen. The effect of diethylene glycol monoethyl ether as a vehicle for topical delivery of ivermectin. Veterinary Research Communications. 1995, 19, (4),309-319